missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PP2C beta/PPM1B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30782-100ug
This item is not returnable.
View return policy
Description
PP2C beta/PPM1B Polyclonal antibody specifically detects PP2C beta/PPM1B in Bovine,Human,Mouse,Rat samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| PP2C beta/PPM1B | |
| Polyclonal | |
| Western Blot : 5-10 μg/mL, Immunoprecipitation : 5-10 μg/mL | |
| EC 3.1.3.16, MGC21657, PP2C-beta, PP2C-beta-X, PP2CBmagnesium-dependent, beta isoform, protein phosphatase 1B, Protein phosphatase 2C isoform beta, protein phosphatase 2C-like protein, protein phosphatase, Mg2+/Mn2+ dependent, 1B | |
| The purified peptide conjugated to KLH. | |
| 100 μg | |
| Stem Cell Markers | |
| 5495 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Bovine, Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction