missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PP2A alpha Antibody (H5D12) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£171.00 - £478.00
Specifications
| Antigen | PP2A alpha |
|---|---|
| Clone | H5D12 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18123843
|
Novus Biologicals
NBP2-29818-0.025ML |
0.025 mL |
£171.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18110804
|
Novus Biologicals
NBP2-29818 |
0.1 mL |
£478.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PP2A alpha Monoclonal specifically detects PP2A alpha in Human samples. It is validated for Western Blot.Specifications
| PP2A alpha | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| EC 3.1.3.16, PP2A-alpha, PP2Ac, PP2CA, PP2Calpha, protein phosphatase 2 (formerly 2A), catalytic subunit, alpha isoform, protein phosphatase 2, catalytic subunit, alpha isozyme, protein phosphatase 2A catalytic subunit, alpha isoform, Replication protein C, RP-C, serine/threonine protein phosphatase 2A, catalytic subunit, alpha isoform, serine/threonine-protein phosphatase 2A catalytic subunit alpha isoform | |
| PPP2CA | |
| IgG1 | |
| Affinity Purified | |
| Human PP2A, B subunit, B56 delta. This monoclonal antibody is the delta isoform for the B56 family of protein phosphatase 2A. |
| H5D12 | |
| Western Blot 1:500-1:2000 | |
| Monoclonal | |
| Mouse | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Neuronal Cell Markers, Neurotransmission, Phospho Specific, Signal Transduction | |
| Q14738 | |
| 5515 | |
| GST fusion protein corresponding to the N-terminus of human PP2A, B subunit, B56 delta. | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title