missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PP2A alpha Antibody (H5D12) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-29818-0.025ML
This item is not returnable.
View return policy
Description
PP2A alpha Monoclonal specifically detects PP2A alpha in Human samples. It is validated for Western Blot.
Specifications
| PP2A alpha | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000 | |
| Q14738 | |
| PPP2CA | |
| GST fusion protein corresponding to the N-terminus of human PP2A, B subunit, B56 delta. | |
| 0.025 mL | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Neuronal Cell Markers, Neurotransmission, Phospho Specific, Signal Transduction | |
| 5515 | |
| Human | |
| IgG1 |
| Western Blot | |
| H5D12 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| EC 3.1.3.16, PP2A-alpha, PP2Ac, PP2CA, PP2Calpha, protein phosphatase 2 (formerly 2A), catalytic subunit, alpha isoform, protein phosphatase 2, catalytic subunit, alpha isozyme, protein phosphatase 2A catalytic subunit, alpha isoform, Replication protein C, RP-C, serine/threonine protein phosphatase 2A, catalytic subunit, alpha isoform, serine/threonine-protein phosphatase 2A catalytic subunit alpha isoform | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human PP2A, B subunit, B56 delta. This monoclonal antibody is the delta isoform for the B56 family of protein phosphatase 2A. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction