missing translation for 'onlineSavingsMsg'
Learn More

RETSCH PP 40 Pellet Presses

Brand:  RETSCH 20.750.0005


Code : A0

Additional Details : Weight : 345.00000kg

 View more versions of this product

Product Code. 10100933

  • £28695.00 / Each

Estimated Shipment: 05-10-2022
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Pellet Press
50/60 Hz
Funnel tube with tamper and 20 Cups
345 kg
220/230 V
40mm dia. Aluminum Cups
Polypropylene
Videos
SDS
Documents
Certificates
Special Offers

Special Offers