missing translation for 'onlineSavingsMsg'
Learn More

POU3F3 Antibody (CL15299) [Unconjugated], Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-43658-25ul

 View more versions of this product

Product Code. 30279386

  • £321.00 / 25µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

POU3F3 Monoclonal antibody specifically detects POU3F3 in Human samples. It is validated for Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

POU3F3
Monoclonal
1 mg/mL
Western Blot 1 μg/mL, Immunocytochemistry/ Immunofluorescence 2-10 μg/mL
brain-1, Brain-specific homeobox/POU domain protein 1, Brn-1, BRN1brn-1, Oct-8, Octamer-binding protein 8, Octamer-binding transcription factor 8, OTF-8, OTF8oct-8, POU class 3 homeobox 3, POU domain class 3, transcription factor 3, POU domain, class 3, transcription factor 3
This antibody was generated using a synthetic peptide of P20264, with the exact immunogen sequence remaining proprietary.
25 μL
Primary
Human
Purified
Western Blot, Immunocytochemistry/Immunofluorescence
CL15299
Unconjugated
PBS, pH 7.2, 40% glycerol
Mouse
Protein A purified
RUO
5455
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG2a
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.