missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POU3F3 Antibody (CL15299) [Unconjugated], Novus Biologicals™
Mouse Monoclonal Antibody
£321.00 - £459.00
Specifications
| Antigen | POU3F3 |
|---|---|
| Clone | CL15299 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, Immunocytochemistry/ Immunofluorescence 2-10 μg/mL |
| Applications | Western Blot, Immunocytochemistry/Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30279386
|
Novus Biologicals
NBP3-43658-25ul |
25 μL |
£321.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30279471
|
Novus Biologicals
NBP3-43658-100ul |
100 μL |
£459.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
POU3F3 Monoclonal antibody specifically detects POU3F3 in Human samples. It is validated for Western Blot,Immunocytochemistry/ ImmunofluorescenceSpecifications
| POU3F3 | |
| 1 mg/mL | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human | |
| brain-1, Brain-specific homeobox/POU domain protein 1, Brn-1, BRN1brn-1, Oct-8, Octamer-binding protein 8, Octamer-binding transcription factor 8, OTF-8, OTF8oct-8, POU class 3 homeobox 3, POU domain class 3, transcription factor 3, POU domain, class 3, transcription factor 3 | |
| This antibody was generated using a synthetic peptide of P20264, with the exact immunogen sequence remaining proprietary. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| CL15299 | |
| Western Blot 1 μg/mL, Immunocytochemistry/ Immunofluorescence 2-10 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.2, 40% glycerol | |
| 5455 | |
| IgG2a | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title