missing translation for 'onlineSavingsMsg'
Learn More

Potassium tin(IV) oxide trihydrate, 95%

Product Code. 11321549
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11321549 1 kg 1kg
11311549 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11321549 Supplier Thermo Scientific Alfa Aesar Supplier No. 014015.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12125-03-0 | H6K2O6Sn | 298.95 g/mol

Potassium tin(IV) oxide is a biochemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12125-03-0
Molecular Formula H6K2O6Sn
Molecular Weight (g/mol) 298.95
MDL Number MFCD00150391
InChI Key HTHDWDSBYOUAFF-UHFFFAOYSA-N
Synonym potassium stannate trihydrate, unii-r38y86o4sy, 2k.so3n.3h2o, potassium tin iv oxide trihydrate, dipotassium dioxido oxo tin trihydrate, dipotassium ion oxostannanebis olate trihydrate, dipotassium oxostannanebis olate trihydrate, dipotassium bis oxidanidyl-oxidanylidene-tin trihydrate
PubChem CID 18694711
IUPAC Name dipotassium;dioxido(oxo)tin;trihydrate
SMILES O.O.O.[K+].[K+].[O-][Sn]([O-])=O

Specifications

Melting Point 140°C (decomposition)
Physical Form Crystalline
Quantity 1 kg
Solubility Information Soluble in water.
Formula Weight 298.94 (244.89 Anhydrous)
Percent Purity ≥95%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Potassium tin(IV) oxide trihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.