accessibility menu, dialog, popup

UserName

CAS RN 12125-03-0

H2OH2OH2OOSnOOKK

CAS RN 12125-03-0

IUPAC Name: dipotassium oxostannanebis(olate) trihydrate
Synonyms: dipotassium oxostannanebis(olate) trihydrate
Molecular Weight (g/mol): 298.95
Molecular Formula: H6K2O6Sn
InChi Key: HTHDWDSBYOUAFF-UHFFFAOYSA-N
SMILES: O.O.O.[K+].[K+].[O-][Sn]([O-])=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results