missing translation for 'onlineSavingsMsg'
Learn More

Potassium nitrate, 99%, Thermo Scientific™

£20.40 - £217.00


Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11421888
SDS View Documents
Thermo Scientific Alfa Aesar
A14527.36
£20.40
500g
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
11431888
SDS View Documents
Thermo Scientific Alfa Aesar
A14527.0E
2500g
£77.20
2500g
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
11441888
SDS View Documents
Thermo Scientific Alfa Aesar
A14527.0C
10000g
£217.00
10000g
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Potassium nitrate has a vast variety of applications. Appropriate preparations of potassium nitrate find use in fertilizers, food preservatives, tooth paste, rocket propellants and fireworks. In addition, it is also used as an electrolyte in a salt bridge, and as thermal storage medium in power generation systems. It forms phase change material with silica and alumina nanoparticles. In combination with boron trifluoride it forms an efficient reagent for nitration of aromatic compounds. Adsorption on alumina provides an environmentally benign aromatic nitrating agent.

Solubility
Moderately soluble in water, and insoluble in alcohol. Soluble in glycerol and ammonia.

Notes
It is an oxidizer. Stable under ordinary conditions. Incompatible with reducing agents and alcohols.
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Potassium nitrate

Signal Word
  • Warning

Hazard Category
  • Oxidising solid Category 3

Hazard Statement
  • H272-May intensify fire; oxidiser.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

Supplemental information
  • MIXTURE LIST-Contain: Potassium nitrate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.