missing translation for 'onlineSavingsMsg'
Learn More

Potassium nitrate, 99%, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar A14527.0E


Quantity 2500g

 View more versions of this product

Product Code. 11431888

  • £77.20 / 2500g
 Restricted Item
In Stock
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Potassium nitrate has a vast variety of applications. Appropriate preparations of potassium nitrate find use in fertilizers, food preservatives, tooth paste, rocket propellants and fireworks. In addition, it is also used as an electrolyte in a salt bridge, and as thermal storage medium in power generation systems. It forms phase change material with silica and alumina nanoparticles. In combination with boron trifluoride it forms an efficient reagent for nitration of aromatic compounds. Adsorption on alumina provides an environmentally benign aromatic nitrating agent.

Solubility
Moderately soluble in water, and insoluble in alcohol. Soluble in glycerol and ammonia.

Notes
It is an oxidizer. Stable under ordinary conditions. Incompatible with reducing agents and alcohols.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

7757-79-1
101.102
FGIUAXJPYTZDNR-UHFFFAOYSA-N
24434
potassium;nitrate
KNO3
MFCD00011409
potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras
CHEBI:63043
[N+](=O)([O-])[O-].[K+]

Specifications

7757-79-1
KNO3
UN1486
potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras
FGIUAXJPYTZDNR-UHFFFAOYSA-N
potassium;nitrate
24434
101.11
Odorless
2.109
2500g
MFCD00011409
14,7648
Moderately soluble in water,and insoluble in alcohol. Soluble in glycerol and ammonia.
[N+](=O)([O-])[O-].[K+]
101.102
CHEBI:63043
99%
Hygroscopic
Potassium nitrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Potassium nitrate

Signal Word
  • Warning

Hazard Category
  • Oxidising solid Category 3

Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.

Supplemental information
  • MIXTURE LIST-Contain: Potassium nitrate

SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.