Learn More
Potassium nitrate, 99.999%, (trace metal basis), Thermo Scientific™
£36.00
Chemical Identifiers
| CAS | 7757-79-1 |
|---|---|
| Molecular Formula | KNO3 |
| Molecular Weight (g/mol) | 101.1 |
| MDL Number | MFCD00011409 |
| InChI Key | FGIUAXJPYTZDNR-UHFFFAOYSA-N |
| Synonym | potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras |
| PubChem CID | 24434 |
| ChEBI | CHEBI:63043 |
| IUPAC Name | potassium;nitrate |
| SMILES | [N+](=O)([O-])[O-].[K+] |
| Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
|
10713085
|
Thermo Scientific Acros
193800100 |
10g | Glass bottle |
£37.10 £36.00 / 10g Save £1.10 2% Off |
Estimated Shipment: 07-09-2026
Log in to see stock. |
||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
For ultra trace analysisChemical Identifiers
| KNO3 | |
| MFCD00011409 | |
| potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras | |
| CHEBI:63043 | |
| [N+](=O)([O-])[O-].[K+] |
Specifications
| 7757-79-1 | |
| KNO3 | |
| MFCD00011409 | |
| 15, 7769 | |
| Solubility in water: 320g/L (20°C). Other solubilities: soluble in glycerol,slightly soluble in alcohol | |
| [N+](=O)([O-])[O-].[K+] | |
| 101.1 | |
| CHEBI:63043 | |
| 99.999% | |
| 2.1 |
| 99.999% | |
| KNO3 | |
| 05,561 | |
| potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras | |
| FGIUAXJPYTZDNR-UHFFFAOYSA-N | |
| potassium;nitrate | |
| 24434 | |
| 101.1 | |
| Trace Metal Basis | |
| Potassium nitrate, 99.999% |
Safety and Handling
GHS H Statement:
May intensify fire; oxidizer.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 231-818-8
missing translation for 'rtecsNumber' : TT3700000
missing translation for 'tsca' : TSCA
- Potassium nitrate
Signal Word
- Warning
Hazard Category
- Oxidising solid Category 3
Hazard Statement
- H272-May intensify fire; oxidiser.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
Supplemental information
- MIXTURE LIST-Contain: Potassium nitrate
RUO – Research Use Only
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.