missing translation for 'onlineSavingsMsg'
Learn More

Potassium nitrate, 99.999%, (trace metal basis)

Product Code. 10713085 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10713085 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10713085 Supplier Thermo Scientific Acros Supplier No. 193800100
  • ONLINE EXCLUSIVE PRICE
    £37.10
    £36.00 / 10g
    Save £1.10
    2% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7757-79-1 | KNO3 | 101.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7757-79-1
Molecular Formula KNO3
Molecular Weight (g/mol) 101.10
MDL Number MFCD00011409
InChI Key FGIUAXJPYTZDNR-UHFFFAOYSA-N
Synonym potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras
PubChem CID 24434
ChEBI CHEBI:63043
SMILES [K+].[O-][N+]([O-])=O

Specifications

Boiling Point 400.0°C
Melting Point 334.0°C
Color White
Physical Form Crystals and/or Chunks
Quantity 10 g
Assay Percent Range 99.999%
Linear Formula KNO3
Fieser 05,561
Merck Index 15, 7769
Solubility Information Solubility in water: 320g/L (20°C). Other solubilities: soluble in glycerol, slightly soluble in alcohol
IUPAC Name potassium;nitrate
Formula Weight 101.1
Percent Purity 99.999%
Grade Trace Metal Basis
Packaging Glass bottle
Density 2.1
Chemical Name or Material Potassium nitrate, 99.999%
Show More Show Less
compliance-icons
Product Identifier
  • Potassium nitrate
Signal Word
  • Warning
Hazard Category
  • Oxidising solid Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
Supplemental information
  • MIXTURE LIST-Contain: Potassium nitrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.