missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium nitrate, 99.994% (metals basis), Thermo Scientific™
44.13 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25g
100g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Quantity | 25g |
UN Number | UN1486 |
Merck Index | 14,7648 |
Solubility Information | Freely soluble in water. Slightly soluble in ethanol. |
Formula Weight | 101.11 |
Percent Purity | 99.994% |
Odor | Odorless |
Sensitivity | Hygroscopic |
Density | 2.109 g/mL |
Assay | (metals basis) |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'