Learn More
Potassium nitrate, 99.994% (metals basis), Thermo Scientific™
£72.30 - £235.00
Chemical Identifiers
| CAS | 7757-79-1 |
|---|---|
| Molecular Formula | KNO3 |
| Molecular Weight (g/mol) | 101.102 |
| MDL Number | MFCD00011409 |
| InChI Key | FGIUAXJPYTZDNR-UHFFFAOYSA-N |
| Synonym | potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras |
| PubChem CID | 24434 |
| ChEBI | CHEBI:63043 |
| IUPAC Name | potassium;nitrate |
| SMILES | [N+](=O)([O-])[O-].[K+] |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
11331618
|
Thermo Scientific Alfa Aesar
011008.14 |
25g |
£72.30
25g |
Estimated Shipment: 03-09-2026
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
11321618
|
Thermo Scientific Alfa Aesar
011008.22 |
100g |
£235.00
100g |
In Stock
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| KNO3 | |
| MFCD00011409 | |
| potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras | |
| CHEBI:63043 | |
| [N+](=O)([O-])[O-].[K+] |
Specifications
| 7757-79-1 | |
| MFCD00011409 | |
| 14,7648 | |
| Freely soluble in water. Slightly soluble in ethanol. | |
| [N+](=O)([O-])[O-].[K+] | |
| 101.102 | |
| CHEBI:63043 | |
| 99.994% | |
| Hygroscopic | |
| (metals basis) |
| KNO3 | |
| UN1486 | |
| potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras | |
| FGIUAXJPYTZDNR-UHFFFAOYSA-N | |
| potassium;nitrate | |
| 24434 | |
| 101.11 | |
| Odorless | |
| 2.109 g/mL | |
| Potassium nitrate |
Safety and Handling
GHS H Statement
H272
May intensify fire
oxidizer.
P210-P220-P221-P280-P370+P378q-P501c
H272
missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: POTASSIUM NITRATE
missing translation for 'einecsNumber' : 231-818-8
missing translation for 'rtecsNumber' : TT3700000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.