Learn More
Potassium nitrate, 99.994% (metals basis)
Description
Potassium nitrate has several applications. It is used as an oxidizer, food preservative, fertilizer, and an active ingredient of condensed aerosol fire suppression systems. It is a powerful nitrating agent in sulfuric acid medium. In addition, it also used as an electrolyte in a salt bridge and thermal storage medium in power generation systems. Ternary salts containing potassium nitrate have been reported to improve the heat storage capacity in the molten salts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 400°C (decomposition) |
| Melting Point | 334°C |
| Color | White |
| Physical Form | Crystalline |
| Quantity | 100 g |
| UN Number | UN1486 |
| Merck Index | 14,7648 |
| Solubility Information | Freely soluble in water. Slightly soluble in ethanol. |
| IUPAC Name | potassium nitrate |
| Formula Weight | 101.11 |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.