missing translation for 'onlineSavingsMsg'
Learn More

Potassium niobium oxide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11356987
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11356987 25 g 25g
11346987 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11356987 Supplier Thermo Scientific Alfa Aesar Supplier No. 010843.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12030-85-2
Molecular Formula KNbO3
Molecular Weight (g/mol) 180.00
MDL Number MFCD00049651
InChI Key UKDIAJWKFXFVFG-UHFFFAOYSA-N
Synonym k.nbo3, potassium ion niobiumoylolate, potassium;oxido dioxo niobium, potassium metacolumbate, potassium niobiumoylolate, potassium niobium oxide, puratronic
PubChem CID 23689307
IUPAC Name potassium;oxido(dioxo)niobium
SMILES [K+].[O-][Nb](=O)=O

Specifications

Physical Form Powder
Quantity 25 g
Solubility Information Slightly soluble in water.
Formula Weight 180.01
Percent Purity 99.999%
Grade Puratronic
Assay (metals basis)
Chemical Name or Material Potassium niobium oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.