accessibility menu, dialog, popup

UserName

KNbOOO

CAS RN 12030-85-2

IUPAC Name: niobium(5+) potassium trioxidandiide
Synonyms: potassium niobate
Molecular Weight (g/mol): 180
Molecular Formula: KNbO3
InChi Key: OMLRJCKUNDGAMS-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[K+].[Nb+5]
Grade 
  • (2)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results