missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POT1 Rabbit anti-Human, Clone: S06-9I1, Novus Biologicals™
Rabbit Monoclonal Antibody
£224.00 - £451.00
Specifications
| Antigen | POT1 |
|---|---|
| Clone | S06-9I1 |
| Applications | Western Blot, Competition Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18367984
|
Novus Biologicals
NBP3-15080-25UL |
25 μg |
£224.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18300893
|
Bio-Techne
NBP3-15080-100UL |
100 μg |
£451.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
POT1 Monoclonal antibody specifically detects POT1 in Human samples. It is validated for Western Blot, Antibody CompetitionSpecifications
| POT1 | |
| Western Blot, Competition Assay | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Cycle and Replication, Chromatin Research, DNA replication Transcription Translation and Splicing | |
| 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
| 25913 | |
| IgG | |
| Affinity-purified |
| S06-9I1 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| DKFZp586D211, hPot1, POT1 protection of telomeres 1 homolog, POT1-like telomere end-binding protein, protection of telomeres 1 homolog (S. pombe), protection of telomeres protein 1 | |
| A synthetic peptide of human POT1 (Uniprot#: Q9NUX5) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title