missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POT1 Rabbit anti-Human, Clone: S06-9I1, Novus Biologicals™
Description
POT1 Monoclonal antibody specifically detects POT1 in Human samples. It is validated for Western Blot, Antibody Competition
Specifications
Specifications
| Antigen | POT1 |
| Applications | Western Blot, Competition Assay |
| Classification | Monoclonal |
| Clone | S06-9I1 |
| Conjugate | Unconjugated |
| Formulation | 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide |
| Gene Alias | DKFZp586D211, hPot1, POT1 protection of telomeres 1 homolog, POT1-like telomere end-binding protein, protection of telomeres 1 homolog (S. pombe), protection of telomeres protein 1 |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide of human POT1 (Uniprot#: Q9NUX5) |
| Purification Method | Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?