missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POT1 Rabbit anti-Human, Clone: S06-9I1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Bio-Techne NBP3-15080-100UL
This item is not returnable.
View return policy
Description
POT1 Monoclonal antibody specifically detects POT1 in Human samples. It is validated for Western Blot, Antibody Competition
Specifications
| POT1 | |
| Monoclonal | |
| Unconjugated | |
| DKFZp586D211, hPot1, POT1 protection of telomeres 1 homolog, POT1-like telomere end-binding protein, protection of telomeres 1 homolog (S. pombe), protection of telomeres protein 1 | |
| A synthetic peptide of human POT1 (Uniprot#: Q9NUX5) | |
| 100 μg | |
| Cancer, Cell Cycle and Replication, Chromatin Research, DNA replication Transcription Translation and Splicing | |
| 25913 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Competition Assay | |
| S06-9I1 | |
| 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
| Rabbit | |
| Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction