missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POT1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 11 publications
Brand: Novus Biologicals NB500-176SS
This item is not returnable.
View return policy
Description
POT1 Polyclonal specifically detects POT1 in Human samples. It is validated for Western Blot.
Specifications
| POT1 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide | |
| DKFZp586D211, hPot1, POT1 protection of telomeres 1 homolog, POT1-like telomere end-binding protein, protection of telomeres 1 homolog (S. pombe), protection of telomeres protein 1 | |
| Rabbit | |
| 71.4 kDa | |
| 0.025 mL | |
| Cancer, Cell Cycle and Replication, Chromatin Research, DNA replication Transcription Translation and Splicing | |
| 25913 | |
| Human | |
| IgG |
| Western Blot | |
| 1.7 mg/mL | |
| Western Blot 1:500-1:2000 (ECL) | |
| Q9NUX5 | |
| POT1 | |
| A synthetic peptide made to an internal portion of the human POT1 protein sequence (between residues 200-300). [UniProt# Q9NUX5] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Immunogen sequence has 95% homology to monkey, 82% homology to rat, and 72% homology to mouse. | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction