missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POT1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 11 publications
£188.00 - £426.00
Specifications
| Antigen | POT1 |
|---|---|
| Concentration | 1.7 mg/mL |
| Dilution | Western Blot 1:500-1:2000 (ECL) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18070796
|
Novus Biologicals
NB500-176SS |
0.025 mL |
£188.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18781674
|
Novus Biologicals
NB500-176 |
£426.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
POT1 Polyclonal specifically detects POT1 in Human samples. It is validated for Western Blot.Specifications
| POT1 | |
| Western Blot 1:500-1:2000 (ECL) | |
| Polyclonal | |
| Rabbit | |
| Cancer, Cell Cycle and Replication, Chromatin Research, DNA replication Transcription Translation and Splicing | |
| Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide | |
| DKFZp586D211, hPot1, POT1 protection of telomeres 1 homolog, POT1-like telomere end-binding protein, protection of telomeres 1 homolog (S. pombe), protection of telomeres protein 1 | |
| POT1 | |
| IgG | |
| Affinity Purified | |
| Human. Immunogen sequence has 95% homology to monkey, 82% homology to rat, and 72% homology to mouse. |
| 1.7 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| Q9NUX5 | |
| 25913 | |
| A synthetic peptide made to an internal portion of the human POT1 protein sequence (between residues 200-300). [UniProt# Q9NUX5] | |
| Primary | |
| Store at 4C. Do not freeze. | |
| 71.4 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title