Learn More
Description
POROS CaptureSelect FibXL Affinity Matrix has specifically been designed for the purification of human fibrinogen from recombinant or plasma source.
Features of this affinity matrix include:
- Fibrinogen purification from human plasma and recombinant sources
- Efficient purification of fibrinogen with high yields and retained activity
- Excellent scalability
- Non-animal derived
- High flow rates
POROS CaptureSelect FibXL Affinity Matrix is an affinity matrix that binds to fibrinogen with high selectivity and yield, enabling a robust, fast, and efficient purification process with excellent purity obtained in one step.
Free of animal components
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).
Main characteristics
- Matrix: cross-linked poly(styrene-divinylbenzene)
- Average particle size: 50 μm
- Ligand: POROS CaptureSelect FibXL affinity ligand
- Ligand coupling method: epoxide
- Binding capacity: approximately 15 g/L resin (determined with plasma derived fibrinogen)
- Elution conditions: acidic elution with 0.1 M glycine pH 3.0; neutral pH elution with 20 mM Tris, 1.0 M MgCl2 and 40% (v/v) propyleneglycol
- Mechanical resistance: 100 bar (1450 psi, 10 MPa)
- Formulation buffer: 20 % (v/v) ethanol
Specifications
Specifications
| Binding Capacity | 15 g/L |
| Matrix | Cross-linked poly(styrene-divinylbenzene) |
| Content And Storage | Store in refrigerator (2°C to 8°C). |
| Shipping Condition | Approved for shipment at Room Temperature or on Wet Ice |
| Particle Size | 50 μm |
| Form | Liquid Suspension |
| Product Line | CaptureSelect |
| Quantity | 50 mL |
| Type | Affinity Matrix |
| Ligand Type | FibXL affinity ligand |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.