missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ POROS™ CaptureSelect™ BacuClear Affinity Matrix

Product Code. p-7203091 Shop All Thermo Scientific Products
Click to view available options
Quantity:
5 mL
10 mL
50 mL
Unit Size:
10mL
50mL
5mL
This item is not returnable. View return policy

Product Code. 18476598

Brand: Thermo Scientific™ 2813742005

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ POROS™ CaptureSelect™ BacuClear Affinity Matrix is specifically designed for the removal of baculovirus contaminants when using baculovirus expression systems (BEVS) for the production of recombinant biomolecules.

POROS CaptureSelect BacuClear Affinity Matrix is specifically designed for the removal of baculovirus contaminants when using baculovirus expression systems (BEVS) for the production of recombinant biomolecules. It is particularly useful when expressing virus and virus-like particles (VLPs), which are similar in size and envelope structure to baculovirus, making discrimination and separation from the co-expressed baculovirus contaminants particularly challenging. The baculovirus-specific CaptureSelect affinity ligand was developed to address this challenge and is coupled to the high-performing POROS matrix. The large pore structure of the POROS beads enables efficient purification of larger biomolecules.

To remove baculovirus impurities, the affinity matrix is operated in flow-through mode. As described by Moleirinho et. al. (1), successful removal of baculovirus particles from influenza VLPs, HCV VLPs, and AAVs was achieved with the affinity ligand identified (BV-2) coupled to the POROS matrix. A dynamic binding capacity for baculovirus particles of 1.6 x 1010 VG/mL affinity matrix was demonstrated with baculovirus removals observed above 70% and VLP recovery yields above 60%.

Features of this affinity matrix include:

  • One-step removal of baculovirus virus particles from virus and VLP containing samples, in flow-through mode
  • Enables a highly selective polishing step for baculovirus removal
  • High specificity, capacity, and retained activity
  • Excellent scalability
  • Non-animal derived

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics:

  • Matrix: cross-linked poly(styrene-divinylbenzene)
  • Average particle size: 50 μm
  • Ligand: POROS CaptureSelect BacuClear affinity ligand
  • Ligand coupling method: epoxide
  • Binding capacity: estimated >1 x 1010 VG/mL of resin
  • Regeneration conditions: acidic stripping condition with 0.5 M acetic acid, pH 2
  • Mechanical resistance: 100 bar (1450 psi, 10 MPa)
  • Formulation buffer: 20% (v/v) ethanol

1. M.G. Moleirinho, R.P. Fernandes, S.B. Carvalho, S. Bezemer, F. Detmers, P Hermans, R.J.S. Silva, P.M. Alves, M.J.T. Carrondo, C. Peixoto. Baculovirus affinity removal in viral-based bioprocesses. Sep. Purif. Technol. 241 (2020)

Specifications

Binding Capacity >1 x 1010 VG/mL of resin (estimated)
Formulation 50% slurry suspension in 20% ethanol
Matrix Cross-linked poly(styrene-divinylbenzene)
Content And Storage Store in refrigerator (2°C to 8°C). Protect from daylight
Max. Operating Pressure 100 bar
Shipping Condition Ships at 2°C to 8°C
Particle Size 50 μm
Form Resin Slurry
Product Line CaptureSelect
Quantity 5 mL
Type Affinity Matrix
Ligand Type BacuClear affinity ligand
Stationary Phase POROS bead
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.