missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ POROS™ CaptureSelect™ AAV8 Affinity Resin

Product Code. 18416828 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 mL
25 mL
50 mL
250 mL
Unit Size:
10mL
250mL
25mL
50mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18416828 250 mL 250mL
18496518 10 mL 10mL
18446678 25 mL 25mL
18476608 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 18416828 Supplier Thermo Scientific™ Supplier No. A30792

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ POROS™ CaptureSelect™ AAV8 high performance affinity resin is engineered to address the high selectivity and capacity requirements needed for the purification of viral vectors used for gene therapy applications and is specifically intended for the purification of adeno-associated virus type 8 (AAV8).

POROS CaptureSelect AAV8 high performance affinity resin is engineered to address the high selectivity and capacity requirements needed for the purification of viral vectors used for gene therapy applications and is specifically intended for the purification of adeno-associated virus type 8 (AAV8).

Features of POROS CaptureSelect AAV8 resin include:

  • One-step AAV8 purification from crude material with high purity and yield
  • High specificity and capacity, which helps reduce the process volume significantly for subsequent steps and maximizes yield
  • Low operating back pressure and linear pressure versus flow responses driving flexible scalability
  • Superior capacity maintained at high flow rates
  • Rigid polymeric bead with covalent surface chemistry for easier handling and packing, with superior physical and chemical stability enabling a more robust downstream purification process
  • Non-animal derived—the ligand is a 13-kDa single-domain fragment that comprises the 3 CDRs that form the antigen-binding domain and is efficiently produced by the yeast Saccharomyces cerevisiae in a production process free of animal components.

POROS CaptureSelect AAV8 affinity resin specifically binds AAV8 and enables a robust, fast, and efficient purification process with excellent purity and yield obtained in one step. The use of this affinity resin will enable significant improvement to downstream processing by reducing the purification steps and maximizing productivity, for scalability and processing consistency.

Main characteristics:

  • Matrix: cross-linked poly(styrene-divinylbenzene)
  • Average particle size: 50 μm
  • Ligand: POROS CaptureSelect AAV8 affinity ligand
  • Ligand coupling method: carbonyldiimidazole (CDI)
  • Binding capacity: >1 x 1013 genome copies/mL of resin
  • Elution conditions: acidic elution with 0.1 M glycine, pH 3.0
  • Mechanical resistance: 100 bar (1,450 psi; 10 MPa)
  • Formulation buffer: 18% (v/v) ethanol

Specifications

Binding Capacity >1 x 1013 genome copies/mL of resin
Matrix Cross-linked poly(styrene-divinylbenzene)
Content And Storage Store at 2°C to 8°C.
Shipping Condition Wet Ice
Particle Size 50 μm
Form Liquid Suspension
Product Line CaptureSelect, POROS
Quantity 250 mL
Type Resin
Ligand Type AAV8 affinity ligand
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.