missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Porimin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£177.00 - £319.00
Specifications
| Antigen | Porimin |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 1-3 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:10-1:500. Use reported in scientific literature (PMID 16814713) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18030816
|
Novus Biologicals
NB100-56371SS |
0.025 mg |
£177.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18085264
|
Novus Biologicals
NB100-56371 |
0.1 mg |
£319.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Porimin Polyclonal specifically detects Porimin in Human, Mouse, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Porimin | |
| Western Blot 1-3 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:10-1:500. Use reported in scientific literature (PMID 16814713) | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Bovine | |
| Q8N131 | |
| 114908 | |
| This antibody was generated by immunizing rabbits with a synthetic peptide corresponding to amino acids 173-188 SRRGIRYRTIDEHDAI of human Porimin; GenBank no. AAG2164.1. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis | |
| PBS containing 0.05% BSA with 0.05% Sodium Azide | |
| KCT-3, KCT3keratinocytes associated transmembrane protein 3, Keratinocytes-associated transmembrane protein 3, PORIMINporimin, pro oncosis receptor inducing membrane injury, Pro-oncosis receptor inducing membrane injury, serine/threonine-rich receptor, transmembrane protein 123PORMIN | |
| TMEM123 | |
| IgG | |
| Protein G purified | |
| Reacts with Human and bovine. This sequence is 100% conserved in cow, and 93% conserved in rat, mouse, and chicken. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title