missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Porimin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB100-56371SS
This item is not returnable.
View return policy
Description
Porimin Polyclonal specifically detects Porimin in Human, Mouse, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Porimin | |
| Polyclonal | |
| Unconjugated | |
| PBS containing 0.05% BSA with 0.05% Sodium Azide | |
| KCT-3, KCT3keratinocytes associated transmembrane protein 3, Keratinocytes-associated transmembrane protein 3, PORIMINporimin, pro oncosis receptor inducing membrane injury, Pro-oncosis receptor inducing membrane injury, serine/threonine-rich receptor, transmembrane protein 123PORMIN | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| Primary | |
| Reacts with Human and bovine. This sequence is 100% conserved in cow, and 93% conserved in rat, mouse, and chicken. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 1-3 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:10-1:500. Use reported in scientific literature (PMID 16814713) | |
| Q8N131 | |
| TMEM123 | |
| This antibody was generated by immunizing rabbits with a synthetic peptide corresponding to amino acids 173-188 SRRGIRYRTIDEHDAI of human Porimin; GenBank no. AAG2164.1. | |
| 0.025 mg | |
| Apoptosis | |
| 114908 | |
| Human, Bovine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction