Learn More
Haldenwanger™ Porcelain Crucible Lids
Use these lids with porcelain crucibles. Haldenwanger™ Porcelain Crucible Lids are constructed from optimized laboratory porcelain.
Brand:  Haldenwanger™ L 79D/6
Code : 02
Additional Details : Weight : 0.01080kg
 View more versions of this product
Product Code. 10210961
  • £9.75 / Each
Estimated Shipment: 30-05-2024
to see stock.
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
  • Available in a variety of sizes
  • For use with Haldenwanger porcelain crucibles
  • Complies with DIN VDE 0335, type C 100
Specifications
Specifications
Porcelain
45mm
Crucible Lid