Learn More
Haldenwanger™ Porcelain Crucible Lids
Product Code. 10311252
Change view
Click to view available options
Diameter (Metric) Outer:
30mm
40mm
45mm
60mm
75mm
87mm
92mm
Unit Size:
Each
Pack of 5
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer unitSize
10311252 60mm Each
10210961 45mm Each
10250911 40mm Each
10557301 30mm Each
12807703 87mm Pack of 5
12817703 92mm Pack of 5
12316497 75mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 10311252 Supplier Haldenwanger™ Supplier No. L 79D/4
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use these lids with porcelain crucibles. Haldenwanger™ Porcelain Crucible Lids are constructed from optimized laboratory porcelain.
  • Available in a variety of sizes
  • For use with Haldenwanger porcelain crucibles
  • Complies with DIN VDE 0335, type C 100
Specifications
Material Porcelain
Diameter (Metric) Outer 60mm
Product Type Crucible Lid