missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Haldenwanger™ Porcelain Buchner Funnel
Use these funnels for a variety of filtration applications. Haldenwanger™ Porcelain Buchner Funnels are constructed from optimized laboratory porcelain.
Brand: Haldenwanger™ 127C/6A
Code : 02
Additional Details : Weight : 9.10800kg
Description
- Available in a variety of sizes
- Available with perforated fixed filter plate
- Complies with DIN VDE 0335, type C 100
Specifications
Buchner Funnel | |
Porcelain | |
279 mm | |
1Each |
5800 mL | |
300 mm | |
279 mm |