missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | POP1 |
|---|---|
| Concentration | 0.39 mg/mL |
| Applications | Immunocytochemistry, Western Blot, Immunofluorescence, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16899493
|
Proteintech
12029-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16889493
|
Proteintech
12029-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Component of ribonuclease P, a protein complex that generates mature tRNA molecules by cleaving their 5′-ends. Also a component of RNase MRP.Specifications
| POP1 | |
| Immunocytochemistry, Western Blot, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q99575 | |
| 10940, 315045, 67724 | |
| POP1 Fusion Protein Ag2660 | |
| Primary | |
| -20°C | |
| POP1 |
| 0.39 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| hPOP1, KIAA0061, POP1 | |
| Pop1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title