Learn More
POP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12029-1-AP-20UL
 View more versions of this product
Product Code. 16899493
  • £135.00 / 20µL
Description
Component of ribonuclease P, a protein complex that generates mature tRNA molecules by cleaving their 5′-ends. Also a component of RNase MRP.
Specifications
Specifications
POP1
Polyclonal
Unconjugated
POP1
hPOP1, KIAA0061, POP1
Rabbit
Antigen Affinity Chromatography
RUO
10940, 315045, 67724
-20°C
Liquid
Immunocytochemistry, Western Blot, Immunofluorescence, Immunoprecipitation
0.39 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q99575
Pop1
POP1 Fusion Protein Ag2660
20 μL
Primary
Human, Mouse, Rat