missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Chromafil™ Polyvinylidene Difluoride Syringe Filters

For aqueous samples, water-soluble oligomers and polymers like proteins.   Macherey Nagel™ Chromafil™ Polyvinylidene Difluoride Syringe Filters are labeled for method validation and certification.

Brand:  Macherey-Nagel™ 729218

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12406446

  • £111.00 / Pack of 100
Estimated Shipment: 26-06-2024
to see stock.
Add to basket

Description

Description

  • Hydrophilic membrane
  • For aqueous samples, water-soluble oligomers and polymers like proteins
  • Binding capacity for proteins 20 μg per 25 mm filter
Specifications

Specifications

PVDF
∼4.9 cm2
100 Pack
∼80 μL
0.20 μm
Polypropylene (PP)
Syringe Filter
25 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers