missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Chromafil™ Polyvinylidene Difluoride Syringe Filters

For aqueous samples, water-soluble oligomers and polymers like proteins.   Macherey Nagel™ Chromafil™ Polyvinylidene Difluoride Syringe Filters are labeled for method validation and certification.

Brand:  Macherey-Nagel™ 729219.400

Code : NEW

Additional Details : Weight : 0.52000kg

 View more versions of this product

Product Code. 10696094

  • £458.21 / Pack of 400
Estimated Shipment: 28-05-2024
to see stock.
Add to basket

Description

Description

  • Hydrophilic membrane
  • For aqueous samples, water-soluble oligomers and polymers like proteins
  • Binding capacity for proteins 20 μg per 25 mm filter
Specifications

Specifications

PVDF
∼4.9 cm2
400 Pack
∼80 μL
0.45 μm
Polypropylene (PP)
Syringe Filter
25 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers