Learn More
Macherey-Nagel™ Chromafil™ Polyvinylidene Difluoride Syringe Filters
For aqueous samples, water-soluble oligomers and polymers like proteins.   Macherey Nagel™ Chromafil™ Polyvinylidene Difluoride Syringe Filters are labeled for method validation and certification.
Brand:  Macherey-Nagel™ 729039
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10413245
  • £145.00 / Pack of 100
Estimated Shipment: 04-06-2024
to see stock.
Add to basket
Description
Description
  • Hydrophilic membrane
  • For aqueous samples, water-soluble oligomers and polymers like proteins
  • Binding capacity for proteins 20 μg per 25 mm filter
Specifications
Specifications
Top Black, Bottom White
1.0/0.45 μm
Polypropylene (PP)
100 Pack
∼80 μL
PVDF with Glass Fiber Prefilter
∼4.9 cm2
Glass Fibre
Syringe Filter
25 mm