Learn More
Macherey-Nagel™ Polystyrene Divinylbenzene Chromabond LV-HR-X
Product Code. 12824311 Shop All Macherey-Nagel Products
Click to view available options
Quantity:
30 mg
60 mg
200 mg
Unit Size:
Each
Product Code. 12824311
Brand: Macherey-Nagel™ 732132
Only null left
Add to basket
Add to basket
Specifications
Quantity 200 mg
Product Type Chromabond LV-HR-X
Particle Size 45 or 85 μm
Material Polystyrene Divinylbenzene
pH Range 1 to 14
Pore Size 55 - 60 Å
Packaging Quantity 30 Pack