missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Eppendorf LoBind material ensures excellent sample recovery for improved assay results
- Free of surface coating (e.g., silicone) to minimize the risk of sample interference
- Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors
- Available in tube, microplate, and deepwell plate formats for easy-up scaling
- High-contrast Unique OptiTrack™ matrix: up to 30 % faster sample identification and fewer pipetting errors
- RecoverMax™ well design: optimized well geometry for minimal remaining/dead volume and excellent mixing properties
- Raised well rims and a smooth surface ensure reliable sealing in plates
Specifications
Specifications
| Format | Standard |
| Array | 16 x 24 |
| Color | White, Clear Bottom |
| Material | Polypropylene |
| No. of Wells | 384 |
| Well Shape | Conical Bottom (V-well) |
| No. per Pack | 240 |
| Quantity | 240/Pk. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?