Learn More
Eppendorf™ Polypropylene V-Bottom 384 Well Microplate
GREENER_CHOICE
Product Code. 10136573 Shop All Eppendorf Products
Change view
Click to view available options
Color:
Gray, White Bottom, White Bottom
White, Black Bottom, Black Bottom
White, Clear Bottom
Certified:
PCR clean
Sterile
Quantity:
240/Pk.
80/Pk.
Unit Size:
Pack of 240
Pack of 80
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Certified Quantity unitSize
10136573 Gray, White Bottom, White Bottom PCR clean 240/Pk. Pack of 240
10678404 White, Clear Bottom PCR clean 240/Pk. Pack of 240
10668404 White, Clear Bottom PCR clean 80/Pk. Pack of 80
10465075 White, Black Bottom, Black Bottom PCR clean 240/Pk. Pack of 240
10208703 White, Clear Bottom Sterile 80/Pk. Pack of 80
10349923 Gray, White Bottom, White Bottom PCR clean 80/Pk. Pack of 80
10218703 White, Clear Bottom Sterile 240/Pk. Pack of 240
10278743 White, Black Bottom, Black Bottom PCR clean 80/Pk. Pack of 80
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 10136573 Supplier Eppendorf™ Supplier No. 0030625676
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Certified PCR clean
Format Standard
Array 16 x 24
Autoclavable Autoclavable
Color Gray, White Bottom, White Bottom
Material Polypropylene
No. of Wells 384
Well Shape Conical Bottom (V-well)
Quantity 240/Pk.