Learn More
Eppendorf™ Polypropylene V-Bottom 384 Well DNA LoBind Microplate
GREENER_CHOICE
Product Code. 10084393 Shop All Eppendorf Products
Change view
Click to view available options
No. per Pack:
80
240
Unit Size:
Pack of 240
Pack of 80
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. per Pack unitSize
10084393 80 Pack of 80
10524714 240 Pack of 240
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10084393 Supplier Eppendorf™ Supplier No. 0030623304
Only null left
Add to basket
Add to basket
Specifications
Format Standard
Array 16 x 24
Color Clear
Material Polypropylene
No. of Wells 384
Sterility Sterile
Well Shape Conical Bottom (V-well)
No. per Pack 80
Quantity 80/Pk.