Learn More
Kartell™ Polypropylene Universal Tip
Reduce sample retention with a tip that offers high clarity and excellent water-repellent properties. Kartell™ Polypropylene Universal Tip uses precise cerntering to guarantee reliable directionality of the sample when delivered.
Brand: Kartell™ 0974900
Code : NEW
Additional Details : Weight : 0.50000kg
Description
Reduce sample retention with a tip that offers high clarity and excellent water-repellent properties. Kartell™ Polypropylene Universal Tip uses precise cerntering to guarantee reliable directionality of the sample when delivered.
- Manufactured from virgin cadmium-free PP
- High clarity and excellent water-repellent properties designed to keep sample retention within the tip to a minimum
- The precise centering of the orifice at the end of the tip guarantees reliable directionality of the sample when delivered
- These tips are recommended for use with the Kartell™ automatic micropipettes but deliver excellent results even if used with the most other micropipette brands available on the market
- Disposable Kartell tips come in different package types to satisfy a variety of needs as economically as possible
Specifications
2 to 200 μL | |
Autoclavable | |
Refill | |
Refill | |
Kartell™, Gilson™, Brand™, Nichiryo™, Biohit™, Socorex™, Eppendorf™, Hamilton™ |
Yellow | |
CE/IVD | |
Universal | |
960/Pack | |
Polypropylene |