Learn More
Kartell™ Polypropylene Universal Tip
Reduce sample retention with a tip that offers high clarity and excellent water-repellent properties. Kartell™ Polypropylene Universal Tip uses precise cerntering to guarantee reliable directionality of the sample when delivered.
£227.00
Specifications
Volume (Metric) | 2 to 200 μL |
---|---|
Color | Yellow |
Autoclavable | Autoclavable |
Certifications/Compliance | CE/IVD |
Tip Style | Universal |
Description
Reduce sample retention with a tip that offers high clarity and excellent water-repellent properties. Kartell™ Polypropylene Universal Tip uses precise cerntering to guarantee reliable directionality of the sample when delivered.
- Manufactured from virgin cadmium-free PP
- High clarity and excellent water-repellent properties designed to keep sample retention within the tip to a minimum
- The precise centering of the orifice at the end of the tip guarantees reliable directionality of the sample when delivered
- These tips are recommended for use with the Kartell™ automatic micropipettes but deliver excellent results even if used with the most other micropipette brands available on the market
- Disposable Kartell tips come in different package types to satisfy a variety of needs as economically as possible
Specifications
2 to 200 μL | |
Autoclavable | |
Universal |
Yellow | |
CE/IVD | |
Polypropylene |