Learn More
Eppendorf™ Polypropylene U-shaped 384 Well Twin Tec PCR Microplate
GREENER_CHOICE
Product Code. 10131982 Shop All Eppendorf Products
Change view
Click to view available options
Color:
Blue
Clear
Green
Red
Yellow
Unit Size:
Pack of 25
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
10131982 Yellow Pack of 25
10374173 Blue Pack of 25
10573035 Clear Pack of 25
10394423 Green Pack of 25
10562065 Red Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10131982 Supplier Eppendorf™ Supplier No. 0030128516
Only null left
Add to basket
Add to basket
Compatible with automated systems. U-shaped inner well contour for easy identification.
Compatible with automated systems. U-shaped inner well contour for easy identification
  • U-shaped inner well contour for easy identification
  • Minimal bending of skirts
  • Continuous skirt for compatibility with automated systems
  • Stackable
  • 8 holes in the skirt aid plate positioning and removal from the thermal cycler block e.g., with grippers
  • Within SBS footprint recommendations (127.76 ±0.25mm x 85.47 ±0.25mm)
  • Suitable for all common 384 well thermocyclers
  • 45μL max well volume
TRUSTED_SUSTAINABILITY
Specifications
Barcode Includes Barcode
Format Standard
Array 16 x 24
Color Yellow
Skirt Style Skirted
Material Polypropylene
No. of Wells 384
Total Volume (Metric) Well 45 μL
Well Shape Round Bottom (U-well)
No. per Pack 25
Plate Compatibility Automated systems
Quantity 25/Pk.