Learn More
Macherey-Nagel™ Polypropylene Reservoir Column
Product Code. 12766333 Shop All Macherey-Nagel Products
Change view
Click to view available options
For Use With (Equipment):
1, 3, 6 mL Chromabond Polypropylene Columns
15, 45, 70 mL CHROMABOND Polypropylene Columns
Quantity:
1 Pack
10 Pack
Unit Size:
Each
Pack of 10
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Quantity Unit Size
12766333 15, 45, 70 mL CHROMABOND Polypropylene Columns 10 Pack Each
10348551 1, 3, 6 mL Chromabond Polypropylene Columns 1 Pack Each
10621502 15, 45, 70 mL CHROMABOND Polypropylene Columns 1 Pack Each
10546701 15, 45, 70 mL CHROMABOND Polypropylene Columns 10 Pack Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12766333 Supplier Macherey-Nagel™ Supplier No. 730389
Estimated Shipment: 20-01-2027