Learn More
Fisherbrand™ Polypropylene Natural Pipet Tip
Make pipetting simple and safer for even the most sensitive applications. Fisherbrand™ Polypropylene Natural Pipet Tips are perfectly fit for manual or electronic, single or multichannel pipets.
£9.40 - £77.55
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
11557452
|
Fisherbrand™
|
1 to 250 μL |
£9.40
Pack of 1000 |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
11597442
|
Fisherbrand™
|
1 to 200 μL |
£39.10
Pack of 1000 |
In Stock
Log in to see stock. |
|||||
11567452
|
Fisherbrand™
|
1 to 300 μL |
£77.55
Pack of 1000 |
N/A | |||||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
11587452
|
Fisherbrand™
|
200 to 1000 μL |
£34.95
Pack of 1000 |
In Stock
Log in to see stock. |
||||
11597452
|
Fisherbrand™
|
250 to 1000 μL |
£16.65
Pack of 1000 |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||||
11507462
|
Fisherbrand™
|
500 to 2500 μL |
£47.35
Pack of 250 |
In Stock
Log in to see stock. |
|||||
Description
- Made of high quality polypropylene
- Tight fit due to the precisely fitting cone
- Optimal handling due to the stable shape
- Autoclavable
- Guaranteed dust-free due to special ionization treatment
- Available pre-sterilized or non-sterile
- Excellent transparency due to high quality surfaces
- Minimal residual wetting due to controlled quality of raw materials
- Excellent volume accuracy due to precise tip orifice
- Practice-oriented tip volumes
- Aeroject™ tips with special filters made of hydrophobic PE-HD as
- Aerosol barrier and for protection against contamination
- Suitable for all commonly used pipet types
Specifications
Natural | |
Autoclavable | |
Polypropylene |