Learn More
Fisherbrand™ Polypropylene Natural Pipet Tip
Make pipetting simple and safer for even the most sensitive applications. Fisherbrand™ Polypropylene Natural Pipet Tips are perfectly fit for manual or electronic, single or multichannel pipets.
Brand:  Fisherbrand™
Code : 41
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11567452
  • £77.55 / Pack of 1000
Currently Unavailable: We have no information when this product will be available
View alternative products
Description
Description
  • Made of high quality polypropylene
  • Tight fit due to the precisely fitting cone
  • Optimal handling due to the stable shape
  • Autoclavable
  • Guaranteed dust-free due to special ionization treatment
  • Available pre-sterilized or non-sterile
  • Excellent transparency due to high quality surfaces
  • Minimal residual wetting due to controlled quality of raw materials
  • Excellent volume accuracy due to precise tip orifice
  • Practice-oriented tip volumes
  • Aeroject™ tips with special filters made of hydrophobic PE-HD as
  • Aerosol barrier and for protection against contamination
  • Suitable for all commonly used pipet types
Specifications
Specifications
1 to 300 μL
Autoclavable
Polypropylene
Natural
Yes