missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Tubes™ 3810X
GREENER_CHOICE

Product Code. 10277391 Shop All Eppendorf Products
Change view
Click to view available options
Color:
Blue
Clear
Green
Red
Yellow
Purity or Quality Grade:
Eppendorf Quality
PCR Clean
Unit Size:
Pack of 1000
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Purity or Quality Grade unitSize
10277391 Blue Eppendorf Quality Pack of 1000
10154671 Clear PCR Clean Pack of 1000
10451043 Clear Eppendorf Quality Pack of 1000
10758052 Green Eppendorf Quality Pack of 1000
10596901 Yellow Eppendorf Quality Pack of 1000
10649781 Red Eppendorf Quality Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 10277391 missing translation for 'mfr' Eppendorf™ missing translation for 'supplierNo' 0030125177
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

The easy-to-open, securely closing, Eppendorf Tubes™ 3810X offers comfort and dependability for sample preparation, centrifugation, and storage. One vessel for all applications in molecular biology, chemistry, and biochemistry. Safer than ever: designed for centrifugation up to 30,000 x g.

Use these tubes for all applications in molecular biology, clinical chemistry and biochemistry. Eppendorf™ 3810X Polypropylene Microcentrifuge Tube offers comfort and dependability for sample preparation, centrifugation and storage.

  • Tube lid is easy to open and close for ergonomic operation.
  • Polypropylene provides high resistance to chemicals, mechanical stress, and temperature extremes.
  • g-Safe™: Exceptional centrifugation stability up to 30,000 × g prevents sample loss due to tube breakage and provides extra safety when working with hazardous samples.
  • Manufactured without the use of slip agents, plasticizers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results.
  • Graduation for visual control of the pipetted volume.
  • Frosted lid and surface on the side for easy labeling.
  • Ensured functionality from -86°C to 100°C.
  • Autoclavable when open (121°C, 20 min).
  • Available in Eppendorf Quality and lot-certified PCR clean purity grade.
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Capacity (Metric) 1.5 mL
Color Blue
Material Polypropylene
No. per Pack 1000 Pk.
Temperature Range (English) -122.8°F to +212°F
Temperature Range (Metric) -86°C to +100°C
Sterility Non-sterile
Purity or Quality Grade Eppendorf Quality
Graduated Yes
Capacity (English) 0.05 oz.
Closure Type Flat Frost Lid
Fit for Purpose Certified Quality
Bottom Shape Conical
Max. RCF 30,000 x g
Shape Cylindrical
Product Type Microcentrifuge Tube
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.