missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Duran™ Polypropylene Membrane Screw Caps
Shop All DWK Life Sciences Products
Click to view available options
Thread Size (Metric):
GL 25
GL 32
Unit Size:
Pack of 5
Specifications
Specifications
| Color | Blue |
| Material | Polypropylene |
| Quantity | 5 Pk. |
| Thread Size (Metric) | GL 32 |
| Height (Metric) | 24 mm |
| Product Type | Membrane Screw Cap |
| For Use With (Equipment) | Pressure Equalization |
| Diameter (Metric) | 41 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction