Learn More
Eppendorf™ Polypropylene F-Bottom 384 Well Microplate
GREENER_CHOICE
Product Code. 10095043 Shop All Eppendorf Products
Change view
Click to view available options
Color:
White, Clear Bottom
Sterility:
Non-sterile
Sterile
Quantity:
240/Pk.
80/Pk.
Unit Size:
Pack of 240
Pack of 80
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Sterility Quantity unitSize
10095043 White, Clear Bottom Sterile 240/Pk. Pack of 240
10065283 White, Clear Bottom Non-sterile 80/Pk. Pack of 80
10380554 White, Clear Bottom Non-sterile 240/Pk. Pack of 240
10723814 White, Clear Bottom Sterile 80/Pk. Pack of 80
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10095043 Supplier Eppendorf™ Supplier No. 0030626109
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Format Standard
Array 16 x 24
Autoclavable Autoclavable
Color White, Clear Bottom
Material Polypropylene
No. of Wells 384
Sterility Sterile
Well Shape Flat Bottom (F-well)
Quantity 240/Pk.