missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Polypropylene Caps

Product Code. 10432698 Shop All Corning Products
Change view
Click to view available options
:
Diameter: 45mm; Thread size: GL 45
Product Code.
10432698 Diameter: 45mm; Thread size: GL 45
1 options
This item is not returnable. View return policy

Product Code. 10432698

Brand: Corning™ 401654

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

45 mm polypropylene cap

  • 45 mm polypropylene cap
  • Two 15.24 cm stainless steel tubes
  • Autoclavable
  • Fits Corning glass and plastic vessels with GL45 mm threads

Order Info

DiameterMetricOuter

TRUSTED_SUSTAINABILITY

Specifications

Closure Color Orange
Closure Material Polypropylene
Material Polypropylene
Includes 2 stainless steel tubing ports
Thread Size GL 45
Autoclavable Autoclavable
Sterility Non-sterile
Closure Type Screw Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.