Learn More
VITLAB™ Polypropylene Buchner Funnels
An excellent choice for filtration in organic chemistry labs.
£138.00
Specifications
Material | Polypropylene |
---|---|
Autoclavable | Yes |
Description | Polypropylene filter |
Quantity | 1/Pk. |
Descripción
- Upper and lower parts are detachable for easy cleaning
- Two different volumes and filter diameters are available
Especificaciones
Polypropylene | |
Yes | |
Polypropylene filter | |
1/Pk. |